Anti-CTSD Mouse Monoclonal Antibody [clone: [3F12-1B9]]

51-937
10077-300EA 906.92 CAD
10077-300
Anti-CTSD Mouse Monoclonal Antibody [clone: [3F12-1B9]]
Cathepsin D antibody can be used in immunofluorescence starting at 1:100 - 1:400, Western Blot starting at 2 ?g/mL, and immunohistochemistry starting at 1:50.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CTSD
Clonality: Monoclonal
Clone: [3F12-1B9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR