Anti-EWSR1 Mouse Monoclonal Antibody [clone: [5C10]]

51-944
10079-650EA 906.92 CAD
10079-650
Anti-EWSR1 Mouse Monoclonal Antibody [clone: [5C10]]
EWSR1 antibody can be used in Western Blot, and immunohistochemistry starting at 1:50.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EWSR1
Clonality: Monoclonal
Clone: [5C10]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR