Anti-IRAK2 Mouse Monoclonal Antibody [clone: [6C7]]

Supplier: Prosci

51-964
10077-338EA 1180.43 CAD
10077-338
Anti-IRAK2 Mouse Monoclonal Antibody [clone: [6C7]]
Antibodies
IRAK2 antibody can be used in Western Blot starting at 5 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IRAK2
Clonality: Monoclonal
Clone: [6C7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR