Anti-PIK3C2A Mouse Monoclonal Antibody [clone: [3E7]]

Supplier: Prosci

51-979
10073-544EA 1060.85 CAD
10073-544
Anti-PIK3C2A Mouse Monoclonal Antibody [clone: [3E7]]
Antibodies
PIK3C2A antibody can be used in Western Blot starting at 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store PIK3C2A antibody at -20 °C or lower. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: PIK3C2A
Clonality: Monoclonal
Clone: [3E7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR