Order Entry
ContactUsLinkComponent
Anti-ISL1 Mouse Monoclonal Antibody [clone: [1H9]]
Anti-ISL1 Mouse Monoclonal Antibody [clone: [1H9]]
Catalog #: 10073-552
Supplier:  Prosci
Anti-ISL1 Mouse Monoclonal Antibody [clone: [1H9]]
Catalog #: 10073-552
Supplier:  Prosci
Price
$1,241.37
Log in for availability
Unit of Measure Each (50µl)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ISL LIM homeobox 1
  • Clonality:
    Monoclonal
  • Clone:
    [1H9]
  • Gene ID:
    P61371
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Antigen symbol:
    ISL1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Format:
    Ascites
  • Form:
    Liquid
  • Antigen synonyms:
    ISL1
  • Storage buffer:
    0.03% sodium azide
  • Storage temperature:
    Short term: 4 °C. Long term: Store at –20 °C. Avoid freeze-thaw cycles.
  • Shipping temperature:
    4 °C
  • Immunogen:
    Purified recombinant fragment of human ISL1 expressed in E. coli.
  • Purification:
    Ascites
  • Cat. no.:
    10073-552

Specifications

About this item

Islet 1 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 3-5 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ISL1
Clonality: Monoclonal
Clone: [1H9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Customers Who Bought This Also Bought