Anti-F10 Sheep Polyclonal Antibody

Supplier: Prosci

51-988
10073-562EA 1011.8 CAD
10073-562
Anti-F10 Sheep Polyclonal Antibody
Antibodies
Factor 10 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

50% glycerol

Factor 10 antibody may be stored at 4 °C for short-term only. As with all antibodies avoid freeze/thaw cycles. Store at -20 °C for long term. Aliquots are stable for at least 12 months

Type: Primary
Antigen: F10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR