Anti-ZNF654 Mouse Monoclonal Antibody [clone: [NKI/C3]]

Supplier: Prosci

51-996
10073-578EA 1011.8 CAD
10073-578
Anti-ZNF654 Mouse Monoclonal Antibody [clone: [NKI/C3]]
Antibodies
ZNF654 antibody can be used in ELISA, Western Blot starting at 1:250 - 1:1000, and immunohistochemistry starting at 10 ?g/mL.

Ascites, 50 mM PBS at, pH 7.4, 1% BSA, 15 mM sodium azide

ZNF654 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: znf654
Clonality: Monoclonal
Clone: [NKI/C3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR