Anti-GFP Chicken Polyclonal Antibody

Supplier: Prosci

54-195
10068-080EA 567.24 CAD
10068-080
Anti-GFP Chicken Polyclonal Antibody
Antibodies
GFP antibody can be used at 1:2000 to 1:5000 for both immunoblot and ELISA testing. User needs to try out the titrations for optimization in IHC and ICC.

PBS Buffer containing 0.02% sodium azide.

2-8°C for one week and -20°C as an undiluted liquid for up to two years.

Type: Primary
Antigen: gfp
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: GFP
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR