Anti-Threonine Rabbit Polyclonal Antibody

Supplier: Prosci

54-208
10068-052EA 936.95 CAD
10068-052
Anti-Threonine Rabbit Polyclonal Antibody
Antibodies
Phospho-Threonine antibody can be used at 1:500 to 1:1,000 for ELISA; 1:200-1:1,000 for Western Blot testing and 1ug/mg sample for IP. Customer using ECL system may need to further dilute the Abs. User needs to try out the titration for optimal concentration.

PBS Buffer containing 0.02% sodium azide.

2-8°C for one week and -20°C as an undiluted liquid for up to two years.

Type: Primary
Antigen: Phospho Threonine Antibody
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR