The Transcription factor EB (TFEB) is a member in the basic helix-loop-helix leucine zipper superfamily of transcription factors that is translocated in a subset of renal tumors (1,2). Recent studies have shown that lysosomal biogenesis is regulated by TFEB (3), which is in turn regulated by the mammalian target of rapamycin (mTOR) complex 1. Other evidence suggests that TFEB coordinates the major steps of the autophagic pathway by driving the expression of autophagy and lysosomal genes.
- TFEB antibody can be used for detection of TFEB by immunohistochemistry at 2.5 μg/ml.
TFEB antibody is affinity chromatography purified via peptide column.
TFEB antibody is supplied in PBS containing 0.02% sodium azide.
TFEB antibody can be stored at 4°C for three months and -20°C, stable for up to one year.
Type: Primary
Antigen: TFEB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human