Anti-CHGA Mouse Monoclonal Antibody [clone: [LK2H10]]

70-004
10073-592EA 835.8 CAD
10073-592
Anti-CHGA Mouse Monoclonal Antibody [clone: [LK2H10]]
Chromogranin A antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

Tris/HCl buffer, 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: [LK2H10]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR