Order Entry
ContactUsLinkComponent
Anti-Troponin C type 1 Mouse Monoclonal Antibody
Anti-Troponin C type 1 Mouse Monoclonal Antibody
Catalog #: 10073-604
Supplier:  Avantor
Anti-Troponin C type 1 Mouse Monoclonal Antibody
Catalog #: 10073-604
Supplier:  Avantor
Discontinued
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    troponin C type 1 (slow)
  • Clonality:
    Monoclonal
  • Gene ID:
    P63316
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Antigen symbol:
    TNNC1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    0.05 mg
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    TNNC1
  • Storage buffer:
    PBS, pH 7.4, 0.1% sodium azide.
  • Storage temperature:
    TNNC1 antibody can be stored short term 4 °C. For long term storage aliquot and store at –20 °C. As with all antibodies avoid freeze/thaw cycles.
  • Concentration:
    1 mg/mL
  • Shipping temperature:
    4 °C
  • Immunogen:
    Human cardiac Troponin C.
  • Purification:
    Protein A Column
  • Cat. no.:
    10073-604

Specifications

About this item

TNNC1 antibody can be used in Western Blot starting at 5 - 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TNNC1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Customers Who Bought This Also Bought