Anti-OSR1 Mouse Monoclonal Antibody

70-024
10073-626EA 910.74 CAD
10073-626
Anti-OSR1 Mouse Monoclonal Antibody
OSR1 antibody can be used in Western Blot starting at 1 - 2 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: OSR1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR