Anti-DES Mouse Monoclonal Antibody [clone: [D9]]

70-049
10073-658EA 835.8 CAD
10073-658
Anti-DES Mouse Monoclonal Antibody [clone: [D9]]
DES Monoclonal Antibody can be used for immunohistochemistry (tested on frozen tissues) starting at 1:25 - 1:200, immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL and Western Blot starting at 1:100 - 1:1000.

PBS containing 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DES
Clonality: Monoclonal
Clone: [D9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Mouse, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR