Anti-ACTA1 Mouse Monoclonal Antibody [clone: [3B3]]

Supplier: Prosci

70-064
10079-672EA 1078.68 CAD
10079-672
Anti-ACTA1 Mouse Monoclonal Antibody [clone: [3B3]]
Antibodies
ACTA1 Monoclonal Antibody can be used for immunohistochemistry (tested on frozen tissues) starting at 1:100 - 1:1000, immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL and Western Blot starting at 1:1000 - 1:5000.

PBS containing 0.09% sodium azide

ACTA1 Monoclonal antibody may be stored at +4°C, avoid freezing

Type: Primary
Antigen: ACTA1
Clonality: Monoclonal
Clone: [3B3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Goat, Human, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR