Anti-PTPN7 Mouse Monoclonal Antibody [clone: [AFPK32]]

Supplier: Prosci

70-065
10079-674EA 1042.19 CAD
10079-674
Anti-PTPN7 Mouse Monoclonal Antibody [clone: [AFPK32]]
Antibodies
PTPN7 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1:200 and Western Blot starting at 1:5000.

HEPES with 0.15M NaCl, 0.01% BSA, 0.03% sodium azide, and 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PTPN7
Clonality: Monoclonal
Clone: [AFPK32]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR