Anti-HP Mouse Monoclonal Antibody [clone: [26E11]]

70-083
10079-710EA 1042.19 CAD
10079-710
Anti-HP Mouse Monoclonal Antibody [clone: [26E11]]
Antibodies
HP Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1:200 and Western Blot starting at 1:500.

HEPES with 0.15M NaCl, 0.01% BSA, 0.03% sodium azide, and 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HP
Clonality: Monoclonal
Clone: [26E11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR