Anti-MET Mouse Monoclonal Antibody [clone: [4AT44]]

70-096
10077-376EA 1042.19 CAD
10077-376
Anti-MET Mouse Monoclonal Antibody [clone: [4AT44]]
Antibodies
MET Monoclonal Antibody can be used for ELISA starting at 1:1000, immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL and Western Blot starting at 1:100 - 1:500.

PBS, 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MET
Clonality: Monoclonal
Clone: [4AT44]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR