Anti-ITGAV Mouse Monoclonal Antibody

70-097
10077-378EA 758.96 CAD
10077-378
Anti-ITGAV Mouse Monoclonal Antibody
ITGAV Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ITGAV
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR