Anti-NEFL Mouse Monoclonal Antibody

70-123
10073-706EA 639.14 CAD
10073-706
Anti-NEFL Mouse Monoclonal Antibody
Neurofilament Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1:50 and Western Blot starting at 1:50.

15 mM sodium azide.

Neurofilament Monoclonal antibody may be stored at 4°C; Avoid freeze-thaw cycles.

Type:
Antigen: NEFL
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Cat, Human, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR