Anti-TLR2 Mouse Monoclonal Antibody

70-127
10073-714EA 758.96 CAD
10073-714
Anti-TLR2 Mouse Monoclonal Antibody
TLR2 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 20 ?g/mL and Western Blot starting at 1:500.

PBS containing 0.05% BSA and 0.05% sodium azide

TLR2 Monoclonal antibody may be stored at 4°C; Avoid freeze-thaw cycles.

Type:
Antigen: TLR2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR