Anti-MAPK8 Mouse Monoclonal Antibody [clone: [3B12]]

Supplier: Prosci

70-146
10073-728EA 1157.94 CAD
10073-728
Anti-MAPK8 Mouse Monoclonal Antibody [clone: [3B12]]
Antibodies
MAPK8 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 5 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAPK8
Clonality: Monoclonal
Clone: [3B12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR