Anti-TNFRSF18 Mouse Monoclonal Antibody [clone: [2H4]]

70-160
10073-740EA 934.11 CAD
10073-740
Anti-TNFRSF18 Mouse Monoclonal Antibody [clone: [2H4]]
TNFRSF18 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 5 ?g/mL and Western Blot starting at 1:500 - 1:1000.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TNFRSF18
Clonality: Monoclonal
Clone: [2H4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR