Anti-MAPK8 Mouse Monoclonal Antibody [clone: [3H2]]

Supplier: Prosci

70-167
10074-970EA 1202.25 CAD
10074-970
Anti-MAPK8 Mouse Monoclonal Antibody [clone: [3H2]]
Antibodies
MAPK8 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 2.5 ?g/mL and Western Blot starting at 1:500 - 1:1000.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAPK8
Clonality: Monoclonal
Clone: [3H2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR