To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:malate dehydrogenase 2, NAD (mitochondrial)
- Clonality:Monoclonal
- Clone:[1G12]
- Gene ID:P40926
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Antigen symbol:MDH2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.05 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:MDH2
- Storage buffer:PBS, pH 7.2
- Storage temperature:MDH2 Monoclonal antibody may be stored short term at 4 °C, long term aliquot and store at –20 °C, avoid freeze thaw cycles.
- Concentration:0.5 mg/mL
- Shipping temperature:4 °C
- Immunogen:Recombinant protein
- Purification:Protein A Column
- Cat. no.:10079-748
Specifications
About this item
MDH2 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL and Western Blot starting at 1:500 - 1:1000.
PBS, pH 7.2
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: MDH2
Clonality: Monoclonal
Clone: [1G12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human