Anti-HLA-DPB1 Mouse Monoclonal Antibody [clone: [TDR31.1]]

Supplier: Prosci

70-203
10073-768EA 1042.19 CAD
10073-768
Anti-HLA-DPB1 Mouse Monoclonal Antibody [clone: [TDR31.1]]
Antibodies
MHC Class II Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL.

50 mM sodium phosphate, pH 7.5, 100 mM potassium chloride, 150 mM NaCl, 0.5 mg/ml Gentamicin sulfate

MHC Class II Monoclonal antibody may be stored at Store at +4°C. Do not freeze.

Type: Primary
Antigen: HLA-DPB1
Clonality: Monoclonal
Clone: [TDR31.1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR