Anti-TF Mouse Monoclonal Antibody

Supplier: Prosci

70-206
10079-766EA 1042.19 CAD
10079-766
Anti-TF Mouse Monoclonal Antibody
Antibodies
TF Monoclonal Antibody can be used for ELISA starting at 1:500000 and immunohistochemistry (tested on parafin embedded tissues) starting at 1:50.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TF
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR