Anti-CHGA Mouse Monoclonal Antibody

70-214
10074-974EA 1042.19 CAD
10074-974
Anti-CHGA Mouse Monoclonal Antibody
Antibodies
CHGA Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1:200 and Western Blot starting at 1: 2000.

HEPES with 0.15M NaCl, 0.01% BSA, 0.03% sodium azide, and 50% glycerol

CHGA Monoclonal antibody may be stored at 4°C; Avoid freeze-thaw cycles.

Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR