Anti-CR2 Mouse Monoclonal Antibody [clone: [Bu32]]

70-247
10073-792EA 835.8 CAD
10073-792
Anti-CR2 Mouse Monoclonal Antibody [clone: [Bu32]]
CR2 Monoclonal Antibody can be used for flow cytometry starting at 1:50 - 1:200 and immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL.

PBS, pH 7.4, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CR2
Clonality: Monoclonal
Clone: [Bu32]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR