Anti-CD79B Rat Monoclonal Antibody [clone: [AT107-2]]

Supplier: Prosci
70-263
10073-824EA 1078.68 CAD
10073-824
Anti-CD79B Rat Monoclonal Antibody [clone: [AT107-2]]
Antibodies
CD79B Monoclonal Antibody can be used for flow cytometry starting at 1:50 - 1:100 and immunohistochemistry (tested on parafin embedded tissues) starting at 2 ?g/mL.

Tris buffered saline, pH 8.0, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD79B
Clonality: Monoclonal
Clone: [AT107-2]
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Dog, Human, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR