Anti-TPH1 Sheep Polyclonal Antibody

70-279
10073-844EA 1042.19 CAD
10073-844
Anti-TPH1 Sheep Polyclonal Antibody
Antibodies
TPH1 antibody can be used for staining frozen immunohistochemistry tissues starting at 1:1000, staining paraffin embeded tissues starting at 1:100, and Western Blot starting at 1:1000.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TPH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human, Mammal, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR