Anti-HIST1H1A Mouse Monoclonal Antibody [clone: [AE-4]]

70-286
10073-856EA 835.8 CAD
10073-856
Anti-HIST1H1A Mouse Monoclonal Antibody [clone: [AE-4]]
Histone H1 antibody can be used for staining paraffin embeded tissues starting at 5 ?g/mL, and Western Blot starting at 1:100.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HIST1H1A
Clonality: Monoclonal
Clone: [AE-4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR