Anti-EPCAM Mouse Monoclonal Antibody [clone: [Ber-EP4]]

70-297
10073-868EA 835.8 CAD
10073-868
Anti-EPCAM Mouse Monoclonal Antibody [clone: [Ber-EP4]]
EPCAM antibody can be used for staining paraffin embeded tissues starting at 1:50.

Antibody is stable for 24 months when stored at 2-8°C.

Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: [Ber-EP4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR