Anti-IL10 Mouse Monoclonal Antibody

70-311
10079-808EA 639.14 CAD
10079-808
Anti-IL10 Mouse Monoclonal Antibody
IL10 antibody can be used for staining paraffin embeded tissues starting at 10 ?g/mL, and Western Blot starting at 0.25 - 0.5 ?g/mL.

Store Lyophilized at room temperature up to 1 month; Reconstituted for up to two weeks at 2-8°C. Aliquot and freeze at -20°C for long term storage. Avoid freeze/thaw cycles.

Type:
Antigen: IL10
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR