Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: [Hyb-K41220A]]

Supplier: Prosci

70-318
10079-822EA 1147.68 CAD
10079-822
Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: [Hyb-K41220A]]
Antibodies
HSP90 antibody can be used for staining paraffin embeded tissues starting at 10 ?g/mL, and Western Blot starting at 1 ?g/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Hsp90aa1
Clonality: Monoclonal
Clone: [Hyb-K41220A]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat, Yeast, S. pombe
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR