Anti-GLI1 Rabbit Polyclonal Antibody

Supplier: Prosci

70-343
10077-416EA 1042.19 CAD
10077-416
Anti-GLI1 Rabbit Polyclonal Antibody
Antibodies
GLI1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 - 10 ?g/mL, and Western Blotting starting at 1 - 3 ?g/mL.

PBS, 0.2% gelatin, 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GLI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR