Anti-JMJD6 Rabbit Polyclonal Antibody

70-353
10077-434EA 835.8 CAD
10077-434
Anti-JMJD6 Rabbit Polyclonal Antibody
JMJD6 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 0.25 - 1 ?g/mL.

PBS, 0.05% BSA, 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: JMJD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Bovine, Chicken, Dog, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Orangutan, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR