Anti-EIF4B Rabbit Polyclonal Antibody

Supplier: Prosci

70-358
10077-444EA 983.75 CAD
10077-444
Anti-EIF4B Rabbit Polyclonal Antibody
Antibodies
EImmunofluorescence4B Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 0.25 - 0.5 ?g/mL.

PBS, 0.05% BSA, 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EIF4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Human, Mouse, Orangutan, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR