Anti-PTGES2 Rabbit Polyclonal Antibody

70-460
10079-968EA 639.14 CAD
10079-968
Anti-PTGES2 Rabbit Polyclonal Antibody
PTGES2 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL, and Western Blotting starting at 1:200.

TBS, pH 7.4, containing BSA, glycerol, and 0.02% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PTGES2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR