Anti-GUCY1A3 Rabbit Polyclonal Antibody

70-462
10079-972EA 639.14 CAD
10079-972
Anti-GUCY1A3 Rabbit Polyclonal Antibody
GUCY1A3 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL, and Western Blotting starting at 1:200.

TBS, pH 7.4, containing 50% glycerol, 0.5 mg/ml BSA, and 0.02% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GUCY1A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bat, Gibbon, Bovine, Chicken, Dog, Xenopus, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR