Anti-PTGER4 Rabbit Polyclonal Antibody

70-469
10079-984EA 639.14 CAD
10079-984
Anti-PTGER4 Rabbit Polyclonal Antibody
PTGER4 / EP4 Antibody can be used for Immunocytochemistry, Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL, and Western Blotting starting at 1:200.

TBS containing 50% glycerol and 0.02% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PTGER4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR