Anti-EPHX2 Rabbit Polyclonal Antibody

70-472
10079-990EA 639.14 CAD
10079-990
Anti-EPHX2 Rabbit Polyclonal Antibody
EPHX2 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL, and Western Blotting starting at 3 ?g/mL.

TBS pH 7.4, containing 50% glycerol, 0.5 mg/ml BSA, and 0.01% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EPHX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Bovine, Gorilla, Horse, Human, Orangutan, Pig, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR