Anti-ENPP2 Rabbit Polyclonal Antibody

70-476
10079-998EA 639.14 CAD
10079-998
Anti-ENPP2 Rabbit Polyclonal Antibody
ENPP2 / Autotaxin Antibody can be used for Immunocytochemistry starting at 2 ?g/mL, and Immunohistochemistry in paraffin embeded tissues starting at 2.5 ?g/mL,

TBS, pH 7.4, 0.5mg/ml BSA, 0.02% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ENPP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR