Anti-OXER1 Rabbit Polyclonal Antibody

Supplier: ProSci

70-478
10080-002EA 1042.19 CAD
10080-002
Anti-OXER1 Rabbit Polyclonal Antibody
Antibodies
OXER1 / GPCR Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL, and Western Blotting starting at 4 ?g/mL.

TBS pH 7.4 containing 50% gylcerol, BSA, and 0.02% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: OXER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR