Anti-VIPR1 Rabbit Polyclonal Antibody

70-531
10073-896EA 639.14 CAD
10073-896
Anti-VIPR1 Rabbit Polyclonal Antibody
VImmunoprecipitationR1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 1:200 - 1:2000.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: VIPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR