Anti-PTK7 Rabbit Polyclonal Antibody

Supplier: ProSci
70-567
10073-968EA 1097.77 CAD
10073-968
Anti-PTK7 Rabbit Polyclonal Antibody
Antibodies
PTK7 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:200, and Western Blotting starting at 1:5000.

50% glycerol and 0.025% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PTK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chimpanzee, Chicken, Dog, Horse, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR