Anti-ACAT1 Rabbit Polyclonal Antibody

Supplier: ProSci

70-608
10077-538EA 1042.19 CAD
10077-538
Anti-ACAT1 Rabbit Polyclonal Antibody
Antibodies
  • ACAT1 Antibody can be used for ELISA starting at 1:20000 - 1:1, and Immunohistochemistry in paraffin embeded tissues starting at 2 μg/mL,

Affinity Purified

PBS, pH 7.4, 0.02% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ACAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR