Anti-IL6R Rabbit Polyclonal Antibody

Supplier: ProSci

70-615
10077-552EA 1042.19 CAD
10077-552
Anti-IL6R Rabbit Polyclonal Antibody
Antibodies
IL6R Antibody can be used for ELISA starting at 1:40000 - 1:1, and Immunohistochemistry in paraffin embeded tissues starting at 2.5 ?g/mL,

PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IL6R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR