Anti-TNK2 Rabbit Polyclonal Antibody

Supplier: ProSci

70-617
10077-556EA 1042.19 CAD
10077-556
Anti-TNK2 Rabbit Polyclonal Antibody
Antibodies
TNK2 / ACK1 Antibody can be used for ELISA starting at 1:16000, and Immunohistochemistry in paraffin embeded tissues starting at 1.25 ?g/mL,

PBS, pH 7.4, 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TNK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR