Anti-HIST1H2AD Rabbit Polyclonal Antibody

Supplier: ProSci
70-633
10075-092EA 758.96 CAD
10075-092
Anti-HIST1H2AD Rabbit Polyclonal Antibody
Histone H2A Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:100, and Western Blotting starting at 1:2000 - 1:10000.

0.05% sodium azide, 38% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HIST1H2AD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR